Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028636.2 | NP_082912.1 | 8, 9 | 1015 | ||
GenBank mRNA | AK036765.1 | - | 51 | |||
AK049370.1 | - | 8, 9 | 1036 | |||
AK153935.1 | - | 8, 9 | 1036 | |||
AK163077.1 | - | 8, 9 | 1022 | |||
AK170642.1 | - | 8, 9 | 1045 | |||
AK171110.1 | - | 9, 10 | 1148 | |||
BC016253.1 | - | 8, 9 | 1015 | 4911979 |