Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028636.2 | NP_082912.1 | 19 | 2243 | ||
GenBank mRNA | AK036765.1 | - | 1279 | |||
AK049370.1 | - | 19 | 2264 | |||
AK153935.1 | - | 19 | 2264 | |||
AK163077.1 | - | 19 | 2245 | |||
AK170642.1 | - | 19 | 2272 | |||
AK171110.1 | - | 20 | 2376 | |||
BC016253.1 | - | 19 | 2243 | 4911979 | ||
BC023038.1 | - | 1 | 14 | |||
BC039913.1 | - | 3 | 408 |