Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028662.2 | NP_082938.1 | 3 | 1254 | ||
GenBank mRNA | AF414190.1 | - | 4 | 927 | ||
AK090329.1 | - | 3 | 1443 | |||
AK144403.1 | - | 4 | 950 | |||
AK152362.1 | - | 4 | 928 | |||
AK154868.1 | - | 4 | 913 | |||
AK155551.1 | - | 4 | 960 | |||
AK167343.1 | - | 3 | 1255 | |||
AK201993.1 | - | 1 | 109 | |||
BC025875.1 | - | 4 | 926 | 5137689 | ||
BC036992.1 | - | 4 | 934 | 5003855 |