Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028665.4 | NP_082941.2 | 7 | 1836 | ||
GenBank mRNA | AK016847.1 | - | 2 | 117 | ||
AK029511.1 | - | 7 | 1838 | |||
AK029888.1 | - | 7 | 1797 | |||
AK033599.1 | - | 7 | 1825 | |||
AK048925.1 | - | 7 | 1730 | |||
AK133339.1 | - | 1797 | ||||
BC030885.1 | - | 7 | 1465 | |||
BC100433.1 | - | 7 | 1872 | |||
BC145456.1 | - | 7 | 954 | 9054017 | ||
BC145457.1 | - | 6 | 818 | |||
BC145458.1 | - | 6 | 818 |