Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D3Ertd751e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099785.1 | NP_001093255.1 | 3 | 296 | ||
NM_001291048.1 | NP_001277977.1 | 3 | 635 | |||
NM_027271.1 | NP_081547.1 | 4 | 413 | |||
NM_028667.3 | NP_082943.2 | 4 | 413 | |||
GenBank mRNA | AK012700.1 | - | 3 | 594 | ||
AK015146.1 | - | 3 | 367 | |||
AK039725.1 | - | 3 | 260 | |||
AK044548.1 | - | 3 | 251 | |||
AK045813.1 | - | 3 | 260 | |||
AK085404.1 | - | 3 | 294 | |||
AK138405.1 | - | 4 | 414 | |||
AK144064.1 | - | 3 | 260 | |||
BC062244.1 | - | 4 | 373 | 1345984 |