Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D3Ertd751e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099785.1 | NP_001093255.1 | 6 | 585 | ||
NM_001291048.1 | NP_001277977.1 | 4 | 672 | |||
NM_027271.1 | NP_081547.1 | 5 | 450 | |||
NM_028667.3 | NP_082943.2 | 7 | 702 | |||
GenBank mRNA | AK012700.1 | - | 4 | 631 | ||
AK015146.1 | - | 6 | 656 | |||
AK039725.1 | - | 6 | 549 | |||
AK044548.1 | - | 4 | 288 | |||
AK045813.1 | - | 6 | 549 | |||
AK085404.1 | - | 6 | 583 | |||
AK138405.1 | - | 8 | 807 | |||
AK144064.1 | - | 6 | 549 | |||
BC062244.1 | - | 5 | 410 | 1345984 |