Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028705.3 | NP_082981.3 | 7 | 942 | ||
GenBank mRNA | AK017531.1 | - | 7 | 915 | ||
AK122192.1 | - | 5 | 511 | |||
BC042574.1 | - | 7 | 954 | 4503584 | ||
BC145978.1 | - | 7 | 944 | 40131281 | ||
BC145980.1 | - | 7 | 944 | 40131283 |