Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289602.1 | NP_001276531.1 | 14, 15 | 2538 | ||
NM_028707.4 | NP_082983.3 | 14, 15 | 2544 | |||
GenBank mRNA | AK018116.1 | - | 9, 10 | 1000 | ||
AK034575.1 | - | 14, 15 | 2314 | |||
AK045529.1 | - | 14, 15 | 2275 | |||
AK053835.1 | - | 14, 15 | 2192 | |||
AK149027.1 | - | 14, 15 | 2274 | |||
BC056437.1 | - | 10, 11 | 1043 | |||
BC062930.1 | - | 13, 14 | 2125 | 6855293 | ||
BC065053.1 | - | 14, 15 | 2312 | 5709231 | ||
BC065055.1 | - | 14, 15 | 2295 | 5720949 | ||
BC066026.1 | - | 14, 15 | 2544 | 5721455 | ||
BC066036.1 | - | 14, 15 | 2301 | 6837266 |