Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028713.1 | NP_082989.1 | 4 | 989 | ||
GenBank mRNA | AK012229.2 | - | 2 | 687 | ||
AK014318.1 | - | 4 | 741 | |||
BC026871.1 | - | 4 | 553 | 3966594 | ||
BC038341.1 | - | 4 | 989 | 4218473 |