Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028715.3 | NP_082991.3 | 15 | 1179 | ||
GenBank mRNA | AA014311.1 | - | 6 | 340 | ||
AK014382.1 | - | 15 | 1164 | |||
AK088957.1 | - | 15 | 1167 | |||
AK137333.1 | - | 15 | 1166 | |||
AK172927.1 | - | 10 | 1690 | |||
BC032207.1 | - | 14 | 1155 | 5323527 |