Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028715.3 | NP_082991.3 | 7, 8 | 612 | ||
GenBank mRNA | AK014382.1 | - | 7, 8 | 597 | ||
AK088957.1 | - | 7, 8 | 596 | |||
AK137333.1 | - | 7, 8 | 599 | |||
AK172927.1 | - | 4, 5 | 483 | |||
BC032207.1 | - | 6, 7 | 588 | 5323527 |