Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079814.1 | NP_001073282.1 | 14 | 1479 | ||
NM_028720.2 | NP_082996.2 | 14 | 1461 | |||
GenBank mRNA | AK014456.1 | - | 14 | 1401 | ||
AK040841.1 | - | 14 | 1411 | |||
AK145277.1 | - | 14 | 1412 | |||
AK150349.1 | - | 14 | 1405 | |||
AK151832.1 | - | 14 | 1427 | |||
AK152586.1 | - | 14 | 1427 | |||
AK152887.1 | - | 14 | 1405 | |||
AK159476.1 | - | 14 | 1415 | |||
AK180672.1 | - | 2 | 204 | |||
AK190428.1 | - | 2 | 283 | |||
AK199843.1 | - | 2 | 204 | |||
AK199856.1 | - | 2 | 205 | |||
AK200495.1 | - | 2 | 204 | |||
AK200607.1 | - | 2 | 205 | |||
BC006893.1 | - | 14 | 1395 | 3599477 |