Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079814.1 | NP_001073282.1 | 5 | 399 | ||
NM_028720.2 | NP_082996.2 | 5 | 399 | |||
GenBank mRNA | AK040841.1 | - | 5 | 349 | ||
AK140812.1 | - | 4 | 1366 | |||
AK145277.1 | - | 5 | 349 | |||
AK150349.1 | - | 5 | 343 | |||
AK151832.1 | - | 5 | 346 | |||
AK152586.1 | - | 5 | 346 | |||
AK152887.1 | - | 5 | 343 | |||
AK159476.1 | - | 5 | 353 | |||
AK181131.1 | - | 3 | 225 | |||
AK181230.1 | - | 3 | 224 | |||
AK186402.1 | - | 3 | 239 | |||
AK213056.1 | - | 2 | 87 | |||
AK214807.1 | - | 3 | 244 | |||
AK215337.1 | - | 3 | 212 | |||
BC006893.1 | - | 5 | 333 | 3599477 |