Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030195.1 | - | 2 | 480 | ||
AK014383.2 | - | 2 | 355 | |||
AK045045.1 | - | 2 | 489 | |||
BC017602.1 | - | 2 | 501 | 4503004 | ||
BC089516.1 | - | 2 | 711 | |||
BC103778.1 | - | 2 | 423 | 6493935 | ||
BC116935.1 | - | 2 | 341 | 40126254 | ||
BC116937.1 | - | 2 | 343 | 40126256 |