Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr42e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028725.3 | NP_083001.1 | 3 | 1425 | ||
GenBank mRNA | AK014586.1 | - | 3 | 1427 | ||
BC018550.1 | - | 1 | 463 | |||
BC038819.1 | - | 3 | 1164 | 3668862 |