Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271414.1 | NP_001258343.1 | 1 | 201 | ||
NM_028728.3 | NP_083004.1 | 1 | 201 | |||
GenBank mRNA | AK014802.2 | - | 1 | 202 | ||
BC150153.1 | - | 1 | 146 | |||
BC150488.1 | - | 1 | 146 | 40141021 |