Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277292.1 | NP_001264221.1 | 17 | 2328 | ||
NM_001277293.1 | NP_001264222.1 | 18 | 2484 | |||
NM_001277294.1 | NP_001264223.1 | 17 | 2185 | |||
NM_001277295.1 | NP_001264224.1 | 8 | 1120 | |||
NM_028736.2 | NP_083012.1 | 18 | 2341 | |||
NM_130891.2 | NP_570961.1 | 16 | 2104 | |||
NM_133442.2 | NP_597699.1 | 16 | 2247 | |||
GenBank mRNA | AB051560.1 | - | 16 | 1981 | ||
AB051561.1 | - | 18 | 2218 | |||
AB051562.1 | - | 16 | 1967 | |||
AK016420.1 | - | 8 | 1118 | |||
AK029845.1 | - | 17 | 2841 | |||
AK029905.1 | - | 8 | 1121 | |||
AK220553.1 | - | 17 | 2042 | |||
AY255674.1 | - | 8 | 936 | |||
AY294283.1 | - | 8 | 917 | |||
BC067398.1 | - | 17 | 2185 | 30096759 | ||
BC072632.1 | - | 18 | 2484 | 6843802 |