Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277292.1 | NP_001264221.1 | 16 | 2176 | ||
NM_001277293.1 | NP_001264222.1 | 17 | 2332 | |||
NM_001277294.1 | NP_001264223.1 | 16 | 2033 | |||
NM_001277295.1 | NP_001264224.1 | 7 | 968 | |||
NM_028736.2 | NP_083012.1 | 17 | 2189 | |||
NM_130891.2 | NP_570961.1 | 15 | 1952 | |||
NM_133442.2 | NP_597699.1 | 15 | 2095 | |||
GenBank mRNA | AB051560.1 | - | 15 | 1829 | ||
AB051561.1 | - | 17 | 2066 | |||
AB051562.1 | - | 15 | 1815 | |||
AK016420.1 | - | 7 | 966 | |||
AK029845.1 | - | 16 | 2689 | |||
AK029905.1 | - | 7 | 969 | |||
AK220553.1 | - | 16 | 1890 | |||
AY255674.1 | - | 7 | 784 | |||
AY294283.1 | - | 7 | 765 | |||
BC067398.1 | - | 16 | 2033 | 30096759 | ||
BC072632.1 | - | 17 | 2332 | 6843802 |