Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159638.1 | NP_001153110.1 | 2 | 252 | ||
NM_028747.2 | NP_083023.1 | 3 | 337 | |||
GenBank mRNA | AK002603.1 | - | 3 | 335 | ||
AK143809.1 | - | 2 | 243 | |||
BC014740.1 | - | 2 | 318 | |||
BC057916.1 | - | 3 | 326 | 5042053 | ||
CB952425.1 | - | 3 | 339 |