Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028747.2 | NP_083023.1 | 2, 3 | 130 | ||
GenBank mRNA | AK002603.1 | - | 2, 3 | 128 | ||
BC014740.1 | - | 1, 2 | 112 | |||
BC057916.1 | - | 2, 3 | 119 | 5042053 | ||
CB952425.1 | - | 2, 3 | 132 |