Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028748.2 | NP_083024.1 | 8 | 1125 | ||
GenBank mRNA | AK002481.1 | - | 8 | 1095 | ||
AK035475.1 | - | 8 | 1137 | |||
AK052776.1 | - | 5 | 676 | |||
AK085411.1 | - | 8 | 1126 | |||
AK086679.1 | - | 8 | 1139 | |||
AY424294.1 | - | 8 | 1095 | |||
BC038908.1 | - | 2 | 281 | |||
BC039662.1 | - | 2 | 281 | |||
BC054855.1 | - | 8 | 1151 | |||
BC100516.1 | - | 7 | 1006 | 30919009 |
Set Membership: |
G-protein coupled receptor |