Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205385.1 | NP_001192314.1 | 13 | 1409 | ||
NM_001205386.1 | NP_001192315.1 | 13 | 1406 | |||
NM_023735.2 | NP_076224.1 | 12 | 1601 | |||
GenBank mRNA | AF307855.1 | - | 3 | 204 | ||
AK004554.1 | - | 12 | 1415 | |||
AK083343.1 | - | 12 | 1419 | |||
AK145536.1 | - | 12 | 1421 | |||
AK150487.1 | - | 12 | 1303 | |||
AK150589.1 | - | 12 | 1379 | |||
AK150991.1 | - | 12 | 1358 | |||
AK152865.1 | - | 12 | 1395 | |||
AK153079.1 | - | 12 | 1422 | |||
AK153150.1 | - | 12 | 1396 | |||
AK153204.1 | - | 12 | 1302 | |||
AK159135.1 | - | 12 | 1388 | |||
AK159668.1 | - | 12 | 1313 | |||
AK159775.1 | - | 12 | 1400 | |||
AK160047.1 | - | 12 | 1261 | |||
AK166529.1 | - | 13 | 1353 | |||
AK166586.1 | - | 13 | 1353 | |||
AK167148.1 | - | 12 | 1360 | |||
AK167891.1 | - | 12 | 1358 | |||
AK168568.1 | - | 12 | 1288 | |||
AK168771.1 | - | 12 | 1374 | |||
AK177697.1 | - | 2 | 83 | |||
BC005557.1 | - | 12 | 1364 | 3156748 | ||
BC080806.1 | - | 13 | 1367 | 5350752 | ||
BC108386.1 | - | 12 | 1332 | 5051125 |