Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205385.1 | NP_001192314.1 | 5 | 473 | ||
NM_001205386.1 | NP_001192315.1 | 5 | 470 | |||
NM_023735.2 | NP_076224.1 | 4 | 665 | |||
GenBank mRNA | AK004554.1 | - | 4 | 479 | ||
AK083343.1 | - | 4 | 483 | |||
AK145536.1 | - | 4 | 485 | |||
AK147931.1 | - | 4 | 315 | |||
AK150487.1 | - | 4 | 367 | |||
AK150589.1 | - | 4 | 443 | |||
AK150991.1 | - | 4 | 423 | |||
AK152865.1 | - | 4 | 460 | |||
AK153079.1 | - | 4 | 486 | |||
AK153150.1 | - | 4 | 460 | |||
AK153204.1 | - | 4 | 366 | |||
AK159135.1 | - | 4 | 452 | |||
AK159668.1 | - | 4 | 377 | |||
AK159775.1 | - | 4 | 464 | |||
AK160047.1 | - | 4 | 325 | |||
AK166529.1 | - | 5 | 417 | |||
AK166586.1 | - | 5 | 417 | |||
AK167148.1 | - | 4 | 424 | |||
AK167891.1 | - | 4 | 422 | |||
AK168568.1 | - | 4 | 352 | |||
AK168771.1 | - | 4 | 438 | |||
BC005557.1 | - | 4 | 428 | 3156748 | ||
BC080806.1 | - | 5 | 431 | 5350752 | ||
BC108386.1 | - | 4 | 396 | 5051125 |