Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256085.1 | NP_001243014.1 | 3 | 283 | ||
NM_001256086.1 | NP_001243015.1 | 4 | 361 | |||
NM_001256087.1 | NP_001243016.1 | 3 | 415 | |||
NM_028774.3 | NP_083050.1 | 3 | 489 | |||
GenBank mRNA | AK004745.1 | - | 3 | 469 | ||
AK029912.1 | - | 3 | 270 | |||
AK150269.1 | - | 3 | 270 | |||
AK152106.1 | - | 3 | 270 | |||
AY039004.1 | - | 3 | 389 | |||
BC138545.1 | - | 4 | 395 | 8861565 | ||
BY136428.1 | - | 3 | 288 | |||
CX237430.1 | - | 3 | 415 |