Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195025.1 | NP_001181954.1 | 8 | 1254 | ||
NM_028778.4 | NP_083054.1 | 7 | 1230 | |||
GenBank mRNA | AK004737.2 | - | 7 | 1218 | ||
AK033672.1 | - | 7 | 1217 | |||
AK034082.1 | - | 8 | 1246 | |||
BC033302.1 | - | 7 | 1205 | 3663595 | ||
BC046833.1 | - | 8 | 1229 | 6816569 |