Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 19 | 3238 | ||
GenBank mRNA | AB093256.1 | - | 16 | 2897 | ||
AK004841.1 | - | 7 | 786 | |||
AK036540.1 | - | 19 | 3222 | |||
AK037082.1 | - | 4 | 318 | |||
AK145708.1 | - | 18 | 3127 | |||
AK150045.1 | - | 19 | 3198 | |||
AK154146.1 | - | 19 | 3227 | |||
AK154925.1 | - | 19 | 3239 | |||
AK157310.1 | - | 19 | 3248 | |||
AK189746.1 | - | 2 | 168 | |||
BC043323.1 | - | 1 | 79 | |||
BC060069.1 | - | 14 | 2350 | |||
BC072573.1 | - | 17 | 3937 | 5705116 | ||
BC094446.1 | - | 10 | 1818 |