Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 21, 22 | 3538 | ||
GenBank mRNA | AB093256.1 | - | 18, 19 | 3197 | ||
AK004841.1 | - | 9, 10 | 1086 | |||
AK036540.1 | - | 21, 22 | 3522 | |||
AK037082.1 | - | 6, 7 | 618 | |||
AK145708.1 | - | 20, 21 | 3427 | |||
AK150045.1 | - | 21, 22 | 3498 | |||
AK154146.1 | - | 21, 22 | 3527 | |||
AK154925.1 | - | 21, 22 | 3539 | |||
AK157310.1 | - | 21, 22 | 3548 | |||
AK202648.1 | - | 1, 2 | 54 | |||
BC043323.1 | - | 3, 4 | 379 | |||
BC060069.1 | - | 16, 17 | 2650 | |||
BC072573.1 | - | 19, 20 | 4237 | 5705116 | ||
BC094446.1 | - | 12, 13 | 2118 |