Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023738.4 | NP_076227.1 | 20 | 2519 | ||
GenBank mRNA | AF288694.1 | - | 15 | 1781 | ||
AK004894.2 | - | 20 | 2520 | |||
AK054410.1 | - | 19 | 2611 | |||
AK089235.1 | - | 19 | 2412 | |||
AK129009.1 | - | 20 | 2429 | |||
AK157941.1 | - | 2 | 846 | |||
AK170421.1 | - | 19 | 2597 | |||
AK170644.1 | - | 18 | 2978 | |||
AK172467.1 | - | 19 | 2857 | |||
BC037693.1 | - | 18 | 2293 | |||
BC138843.1 | - | 20 | 2436 | 8861865 |