Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023738.4 | NP_076227.1 | 7 | 807 | ||
GenBank mRNA | AF288694.1 | - | 2 | 69 | ||
AK004894.2 | - | 7 | 808 | |||
AK054410.1 | - | 7 | 814 | |||
AK089235.1 | - | 7 | 811 | |||
AK129009.1 | - | 7 | 719 | |||
AK170421.1 | - | 7 | 800 | |||
AK170644.1 | - | 7 | 806 | |||
AK172467.1 | - | 7 | 769 | |||
BC037693.1 | - | 5 | 581 | |||
BC138843.1 | - | 7 | 724 | 8861865 |