Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143885.1 | NP_001137357.1 | 14, 15 | 1951 | ||
NM_001143886.1 | NP_001137358.1 | 13, 14 | 1706 | |||
NM_001244990.1 | NP_001231919.1 | 14, 15 | 1951 | |||
NM_002480.2 | NP_002471.1 | 13, 14 | 2095 | |||
XM_005268887.3 | XP_005268944.1 | 13, 14 | 2094 | |||
XM_011538374.1 | XP_011536676.1 | 14, 15 | 2201 | |||
XM_011538375.1 | XP_011536677.1 | 14, 15 | 2165 | |||
XM_011538376.2 | XP_011536678.1 | 14, 15 | 2201 | |||
XM_011538377.1 | XP_011536679.1 | 13, 14 | 2126 | |||
XM_011538378.2 | XP_011536680.1 | 14, 15 | 2201 | |||
XM_017019323.1 | XP_016874812.1 | 13, 14 | 2048 | |||
XM_017019324.1 | XP_016874813.1 | 12, 13 | 2009 | |||
XM_017019325.1 | XP_016874814.1 | 12, 13 | 1973 | |||
XM_017019330.1 | XP_016874819.1 | 14, 15 | 2201 | |||
GenBank mRNA | AF458589.1 | - | 14, 15 | 1951 | ||
AK302692.1 | - | 13, 14 | 1706 | |||
AK314193.1 | - | 13, 14 | 1835 | |||
BC013941.1 | - | 4, 5 | 461 | |||
BC092481.1 | - | 14, 15 | 1951 | |||
BC111752.1 | - | 13, 14 | 1828 | 40008469 | ||
D87930.1 | - | 13, 14 | 1828 |