Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145827.1 | NP_001139299.1 | 9 | 1192 | ||
NM_028800.3 | NP_083076.3 | 7 | 838 | |||
GenBank mRNA | AB102946.1 | - | 7 | 669 | ||
AK009154.1 | - | 2 | 56 | |||
AK050808.1 | - | 3 | 141 | |||
AK154780.1 | - | 9 | 1183 | |||
AY336057.1 | - | 7 | 823 | |||
BC046981.1 | - | 7 | 797 | 6478265 |