Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028803.4 | NP_083079.1 | 9 | 1327 | ||
GenBank mRNA | AK009815.1 | - | 9 | 1143 | ||
AK050365.1 | - | 9 | 1214 | |||
AK050423.1 | - | 9 | 1213 | |||
AK148186.1 | - | 3 | 255 | |||
BC017541.1 | - | 9 | 1188 | 4526961 |