Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028805.2 | NP_083081.2 | 17 | 2000 | ||
GenBank mRNA | AK010364.1 | - | 17 | 1931 | ||
AK031630.1 | - | 17 | 1912 | |||
AK034103.1 | - | 17 | 1920 | |||
AK037858.1 | - | 2158 | ||||
AK049345.1 | - | 17 | 1914 | |||
AK054173.1 | - | 17 | 1972 | |||
AK087973.1 | - | 17 | 1913 | |||
AK136322.1 | - | 17 | 1921 | |||
BC024500.1 | - | 8 | 807 | |||
BC045200.1 | - | 16 | 1911 | 5322489 |