Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023741.2 | NP_076230.1 | 2, 3 | 269 | ||
GenBank mRNA | AF230921.1 | - | 2, 3 | 255 | ||
AF230922.1 | - | 2, 3 | 258 | |||
AF230923.1 | - | 3, 4 | 471 | |||
AF466150.1 | - | 2, 3 | 270 | |||
AK005568.1 | - | 2, 3 | 275 | |||
AK167369.1 | - | 2, 3 | 276 | |||
BC100326.1 | - | 2, 3 | 263 | 30787396 |