Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007154.3 | NP_001007155.1 | 3 | 314 | ||
NM_001177789.1 | NP_001171260.1 | 3 | 532 | |||
NM_001177790.1 | NP_001171261.1 | 3 | 532 | |||
NM_001177791.1 | NP_001171262.1 | 3 | 366 | |||
NM_028806.2 | NP_083082.1 | 3 | 771 | |||
GenBank mRNA | AJ632220.1 | - | 3 | 329 | ||
AJ632221.1 | - | 3 | 367 | |||
AJ632222.1 | - | 3 | 314 | |||
AJ632223.1 | - | 2 | 204 | |||
AK005136.1 | - | 3 | 336 | |||
AK032242.1 | - | 3 | 538 | |||
AK038449.1 | - | 3 | 282 | |||
AK039213.1 | - | 3 | 321 | |||
AK135144.1 | - | 3 | 772 | |||
AK220554.1 | - | 3 | 532 | |||
AY501387.1 | - | 3 | 478 | |||
BC058621.1 | - | 3 | 687 | 5708186 | ||
BC059869.1 | - | 3 | 550 | 6809310 | ||
BY127674.1 | - | 3 | 315 |