Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321312.1 | NP_001308241.1 | 6 | 1443 | ||
NM_001321313.1 | NP_001308242.1 | 6 | 1229 | |||
NM_001321314.1 | NP_001308243.1 | 5 | 1149 | |||
NM_001321315.1 | NP_001308244.1 | 4 | 1062 | |||
NM_005966.3 | NP_005957.2 | 8 | 1705 | |||
XM_005246582.1 | XP_005246639.1 | 7 | 1378 | |||
XM_005246583.1 | XP_005246640.1 | 7 | 1492 | |||
XM_005246586.1 | XP_005246643.1 | 6 | 1182 | |||
XM_005246587.1 | XP_005246644.1 | 5 | 1095 | |||
XM_011511219.2 | XP_011509521.1 | 7 | 1347 | |||
XM_017004170.1 | XP_016859659.1 | 7 | 1388 | |||
XM_017004171.1 | XP_016859660.1 | 6 | 1555 | |||
XM_017004172.1 | XP_016859661.1 | 6 | 1327 | |||
XM_017004173.1 | XP_016859662.1 | 7 | 1410 | |||
XM_017004174.1 | XP_016859663.1 | 6 | 1226 | |||
XM_017004175.1 | XP_016859664.1 | 5 | 1304 | |||
XM_017004176.1 | XP_016859665.1 | 6 | 1297 | |||
GenBank mRNA | AF045451.1 | - | 5 | 1152 | ||
AF045452.1 | - | 4 | 1065 | |||
AK292446.1 | - | 6 | 1243 | |||
AK296549.1 | - | 4 | 390 | |||
AK313542.1 | - | 5 | 1247 | |||
BC035724.1 | - | 8 | 1594 | 5721972 | ||
U47007.1 | - | 5 | 1149 |