Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253812.1 | NP_001240741.1 | 13 | 1580 | ||
NM_028811.3 | NP_083087.1 | 13 | 1523 | |||
GenBank mRNA | AK012072.1 | - | 13 | 1461 | ||
AK088457.1 | - | 13 | 1544 | |||
AK150058.1 | - | 13 | 1461 | |||
BC026461.1 | - | 11 | 1175 | |||
BC037470.1 | - | 7 | 877 | |||
BC048822.1 | - | 8 | 1028 | |||
BC057453.1 | - | 13 | 1463 | 3969625 |