Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152817.4 | NP_690030.3 | 17, 18 | 2379 | ||
GenBank mRNA | AK012131.1 | - | 17, 18 | 2379 | ||
AK030942.1 | - | 17, 18 | 2380 | |||
AK049324.1 | - | 17, 18 | 2380 | |||
AK084905.1 | - | 1 | 2943 | |||
AK168138.1 | - | 17, 18 | 2370 | |||
BC021912.1 | - | 16, 17 | 2114 | 4503335 |