Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028815.4 | NP_083091.1 | 6 | 650 | ||
GenBank mRNA | AK012971.2 | - | 6 | 635 | ||
AK016556.1 | - | 6 | 725 | |||
AK046486.1 | - | 6 | 1199 | |||
AK154885.1 | - | 6 | 629 | |||
BC067049.1 | - | 6 | 650 | 30363673 |