Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 17 | 2568 | ||
NM_001166548.1 | NP_001160020.1 | 16 | 2500 | |||
NM_001166549.1 | NP_001160021.1 | 16 | 2604 | |||
NM_023743.2 | NP_076232.2 | 17 | 2583 | |||
GenBank mRNA | AB031388.1 | - | 17 | 2404 | ||
AK012082.1 | - | 7 | 987 | |||
AK041598.1 | - | 16 | 2462 | |||
AK088425.1 | - | 12 | 1670 | |||
AK128990.1 | - | 16 | 2602 | |||
AK165678.1 | - | 17 | 2570 | |||
BC033410.1 | - | 17 | 2418 | 5359290 |