Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 6 | 862 | ||
NM_001166548.1 | NP_001160020.1 | 6 | 866 | |||
NM_001166549.1 | NP_001160021.1 | 6 | 970 | |||
NM_023743.2 | NP_076232.2 | 6 | 877 | |||
GenBank mRNA | AB031388.1 | - | 6 | 698 | ||
AK041598.1 | - | 6 | 828 | |||
AK090363.1 | - | 5 | 1355 | |||
AK128990.1 | - | 6 | 968 | |||
AK165678.1 | - | 6 | 865 | |||
AK205705.1 | - | 1 | 43 | |||
BC033410.1 | - | 6 | 712 | 5359290 |