Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033606.2 | NP_001028778.2 | 2 | 581 | ||
NM_028817.3 | NP_083093.2 | 3 | 721 | |||
GenBank mRNA | AK012088.1 | - | 2 | 524 | ||
AK161708.1 | - | 3 | 722 | |||
AK166723.1 | - | 3 | 660 | |||
BC031529.1 | - | 1 | 306 | 4009004 |