Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134077.4 | NP_598838.3 | 6 | 1149 | ||
GenBank mRNA | AK049083.1 | - | 5 | 605 | ||
AK051686.1 | - | 6 | 1125 | |||
AK139077.1 | - | 6 | 1150 | |||
AK172039.1 | - | 6 | 1128 | |||
BC049360.1 | - | 6 | 1125 | |||
BC066051.1 | - | 6 | 1086 |