Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028826.1 | NP_083102.1 | 7 | 912 | ||
GenBank mRNA | AK006043.1 | - | 2 | 103 |