Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286007.1 | NP_001272936.1 | 5 | 845 | ||
NM_001286009.1 | NP_001272938.1 | 5 | 845 | |||
NM_133755.2 | NP_598516.2 | 5 | 772 | |||
GenBank mRNA | AK033157.1 | - | 5 | 562 | ||
AK045084.1 | - | 5 | 561 | |||
AK146771.1 | - | 5 | 767 | |||
AK153511.1 | - | 5 | 693 | |||
AK167924.1 | - | 5 | 667 | |||
BC012519.1 | - | 5 | 700 | 3601589 | ||
BC025582.1 | - | 5 | 661 | 5324981 | ||
BC027034.1 | - | 5 | 756 |