Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028832.3 | NP_083108.1 | 3 | 1184 | ||
GenBank mRNA | AK047778.1 | - | 3 | 1160 | ||
BC046619.1 | - | 2 | 1131 | 6494011 |