Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001565.2 | NP_001001565.1 | 3 | 948 | ||
NM_001001566.3 | NP_001001566.1 | 2 | 805 | |||
GenBank mRNA | AK155424.1 | - | 2 | 465 | ||
AK160734.1 | - | 1 | 1053 | |||
BC030889.1 | - | 2 | 293 | |||
BC057050.1 | - | 3 | 784 | |||
BC060598.1 | - | 3 | 784 | |||
BC071273.1 | - | 2 | 605 | 6334418 |