Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293672.1 | NP_001280601.1 | 5, 6 | 774 | ||
NM_028836.4 | NP_083112.1 | 5, 6 | 774 | |||
GenBank mRNA | AK008791.2 | - | 5, 6 | 749 | ||
BC022594.1 | - | 4, 5 | 535 | |||
DQ100384.1 | - | 5, 6 | 748 |