Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033171.2 | NP_001028343.2 | 3, 4 | 1118 | ||
GenBank mRNA | AK009417.1 | - | 3, 4 | 1119 | ||
AK145064.1 | - | 3, 4 | 1113 | |||
AK162933.1 | - | 4, 5 | 706 | |||
AK169862.1 | - | 4, 5 | 662 |