Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021535.4 | NP_067510.3 | 12 | 1546 | ||
GenBank mRNA | AB044414.1 | - | 12 | 1475 | ||
AK003493.1 | - | 12 | 1533 | |||
AK019174.1 | - | 12 | 1505 | |||
AK145982.1 | - | 12 | 1545 | |||
AK159635.1 | - | 12 | 1533 | |||
AK168813.1 | - | 12 | 1540 | |||
AK212788.1 | - | 3 | 351 | |||
BC057446.1 | - | 12 | 1507 | 3155760 |